Folding precision magnifiers
Folding magnifier, aplanatic, 23 mm
630-8232
Swivelling black metallic magnifier with loop, lens frame made from black painted metal.
- Protective case can serve as a handle
- Provide distortion-free images
- Chrome plated metal
- Ø:23 mm
- Magnification:8×
- Description:Folding magnifier, aplanatic
- Seq:001