Orders LinkContactUsLinkComponent
Binocular magnifiers
Binocular magnifiers
Binocular magnifying lens with 250 mm working distance*
Catalog # 630-8214
Supplier:   ESCHENBACH OPTIK
Binocular magnifiers
Catalog # 630-8214
Supplier:  ESCHENBACH OPTIK
Binocular magnifiers
Binocular magnifiers
Binocular magnifying lens with 250 mm working distance*
Price
191,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Magnifiers with discrete metal frame and black matt finish for use with monocular lens components.
* Shorter working distance due to integrated convergence supporting prisms.
Delivery information: Supplied with a protective box.
Caution: Not suitable for spectacle wearers.
Specifications
  • Magnification:
  • Description:
    Binocular magnifying lens with 250 mm working distance*
  • Seq:
    001