Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Baumé hydrometers

60 - 70, 0,5, 220 mm
  614-0370
Baumé hydrometers
  614-0370
Baumé hydrometers
Baumé hydrometers

60 - 70, 0,5, 220 mm

 
42,50 €
   
1 * 1 ST
 

Baumé hydrometers calibrated at 15 °C.

 
  • Length:
    220 mm
  • Seq:
    27
  • Division:
    0,5
  • Range:
    60 - 70
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.