Order Entry
Belgium
Orders LinkContactUsLinkComponent
Baumé hydrometers

Baumé hydrometers

0 - 10, 0,5, 220 mm
Catalog # 614-0310
Supplier:   Alla France
Baumé hydrometers
Catalog # 614-0310
Supplier:  Alla France
Baumé hydrometers
Baumé hydrometers

0 - 10, 0,5, 220 mm

Price
28,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Baumé hydrometers calibrated at 15 °C.

Specifications
  • Length:
    220 mm
  • Seq:
    02
  • Division:
    0,5
  • Range:
    0 - 10