ICEMACHINE ZBE30-10 + HygPlus, icemachine with production capacity of 30kg/24h, self-contained bin with storage capacity of 10kg, de livered with HygienePlus system, EU-plug 1 * 1 items
Catalog # 474-1200
Supplier: ZIEGRA EISMASCHINEN
Frequently Bought Together