Order Entry
Belgium
Orders LinkContactUsLinkComponent
Forceps, precision, with locking clip

Forceps, precision, with locking clip

Straight, ultra fine tips (tower tip), 5L, with locking clip, 110 mm
Catalog # 232-1522
Supplier:   OUTILS RUBIS
Forceps, precision, with locking clip
Catalog # 232-1522
Supplier:  OUTILS RUBIS
Forceps, precision, with locking clip
Forceps, precision, with locking clip

Straight, ultra fine tips (tower tip), 5L, with locking clip, 110 mm

Price
45,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel. Forceps with very precise, fine tips. For applications that require high precision such as in microscopy, biology and microbiology.

  • Acid-resistant, resistant to hydrofluoric acid and nitric acid
  • 100% antimagnetic
  • Extra fine tips

Documents

Certificates
Search by Lot Number
    Images 360
  • 5L
Specifications
  • Type:
    5L, with locking clip
  • Description:
    Straight, ultra fine tips (tower tip)
  • Length:
    110 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.