Order Entry
Belgium
Orders LinkContactUsLinkComponent
Forceps, precision, with locking clip

Forceps, precision, with locking clip

Straight, ultra fine tips, 3L, with locking clip, 120 mm
Catalog # 232-1521
Supplier:   OUTILS RUBIS
Forceps, precision, with locking clip
Catalog # 232-1521
Supplier:  OUTILS RUBIS
Forceps, precision, with locking clip
Forceps, precision, with locking clip

Straight, ultra fine tips, 3L, with locking clip, 120 mm

Price
51,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel. Forceps with very precise, fine tips. For applications that require high precision such as in microscopy, biology and microbiology.

  • Acid-resistant, resistant to hydrofluoric acid and nitric acid
  • 100% antimagnetic
  • Extra fine tips

Documents

Certificates
Search by Lot Number
    Images 360
  • 5L
Specifications
  • Type:
    3L, with locking clip
  • Description:
    Straight, ultra fine tips
  • Length:
    120 mm
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.