Order Entry
Belgium
Orders LinkContactUsLinkComponent
Stirring spatulas

Stirring spatulas

Standard, 120 mm, 3 mm, Transparent
Catalog # 231-2431
Supplier:   BOETTGER PAUL
Stirring spatulas
Catalog # 231-2431
Supplier:  BOETTGER PAUL
Stirring spatulas
Stirring spatulas

Standard, 120 mm, 3 mm, Transparent

Price
20,40 €
Log in for availability
1 * 500 ST
Product Details & Documents

PS, transparent. Stirring rod for a wide variety of laboratory applications.

  • With or without mixing plate at one end

These spatulas can be used for various applications, such as mixing samples and solutions in test tubes, adding small reagent volumes and releasing blood clots from the side of test tube walls.

Specifications
  • Colour:
    Transparent
  • Description:
    Standard
  • Length:
    120 mm
  • Seq:
    01
  • Blade width:
    3 mm