Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cool Boxes

Cool Boxes

Cooling box ISOTHERM Extreme, 17 l, 365×295×450 mm (ext); 320×195×310 mm (int)
Catalog # 216-6981
Supplier:   CAMPING GAZ
Cool Boxes
Catalog # 216-6981
Supplier:  CAMPING GAZ
Cool Boxes
Cool Boxes

Cooling box ISOTHERM Extreme, 17 l, 365×295×450 mm (ext); 320×195×310 mm (int)

Price
59,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

Blue-white PP. Sturdy, robust design, ideal for storing and transporting temperature-sensitive materials and samples.

  • Box casing and lid fully insulated with CFC-free PU foam
  • A rubber seal ensures the boxes are airtight, a lid with non breakable handle can be securely locked
  • Excellent cooling performance (depending on the model up to 24 hours cooling)
Specifications
  • Capacity:
    17 l
  • Description:
    Cooling box ISOTHERM Extreme
  • Dimensions:
    365×295×450 mm (ext); 320×195×310 mm (int)
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.