Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Racks for test tubes

100 (10×10), Steel, PA coated, 210×210×68 mm
  212-6489
Racks for test tubes
  212-6489
Racks for test tubes
Racks for test tubes

100 (10×10), Steel, PA coated, 210×210×68 mm

 
115,00 €
   
1 * 1 ST
 

Stainless steel, rust-proof, electrolytically or steel polished, polyamide coated.

  • Excellent chemical resistance
  • Excellent thermal resistance (polyamide coating resistant up to 80 °C)
  • Robust, stable construction and easy to clean

Wire racks with second level tier, 21 mm high. Total height 68 mm. Hole size 18×18 mm.

 
  • No. of Holes:
    100 (10×10)
  • Material:
    Steel, PA coated
  • Seq:
    7
  • W×D×H:
    210×210×68 mm