Order Entry
Belgium
Orders LinkContactUsLinkComponent
Conductivity standards

Conductivity standards

Conductivity calibration solution 100 µS/cm, shelf life 2 years, NIST traceable, accuracy ±3%, 300 mL
Catalog # WTWA300578_U
Supplier:   WTW
Conductivity standards
Catalog # WTWA300578_U
Supplier:  WTW
CAS Number:  
Conductivity standards
Conductivity standards

Conductivity calibration solution 100 µS/cm, shelf life 2 years, NIST traceable, accuracy ±3%, 300 mL

CAS Number:  
Price
193,00 €
Log in for availability
1 * 300 mL
Specifications

Specifications

  • Pk:
    300 mL
  • Description:
    Conductivity calibration solution 100 µS/cm, shelf life 2 years, NIST traceable, accuracy ±3%

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.