Orders LinkContactUsLinkComponent
Evaporating flasks
3000 ml, 29/32 NS, coated
Catalog # BUCH025324
Supplier:   BUCHI
Evaporating flasks
Catalog # BUCH025324
Supplier:  BUCHI
Evaporating flasks
Evaporating flasks
3000 ml, 29/32 NS, coated
Price
351,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® rotary evaporators. Centre neck. Pear-shaped with ground joint socket or EL-flange.
Specifications
  • Socket size:
    29/32 NS
  • Type:
    coated
  • Capacity:
    3000 ml
  • Seq:
    22