Orders LinkContactUsLinkComponent
Spider evaporators
Spider with cylindric flasks, SJ24/40, glass, incl. 12 cylindric flasks, SJ14.5/23, 20 ml, For simultaneous distillation from 12×20 ml cylindrical flasks with 24/40 NS Joint
Catalog # BUCH011577
Spider evaporators
Catalog # BUCH011577
Supplier:  
Spider evaporators
Spider evaporators
Spider with cylindric flasks, SJ24/40, glass, incl. 12 cylindric flasks, SJ14.5/23, 20 ml, For simultaneous distillation from 12×20 ml cylindrical flasks with 24/40 NS Joint
Discontinued
Product Details & Documents
Spider evaporators are ideal when users need to concentrate several samples under the same conditions.
  • Models for distillation with evaporating or cylindrical flasks available
  • Parallel distillation saves time
Specifications
  • Description:
    Spider with cylindric flasks, SJ24/40, glass, incl. 12 cylindric flasks, SJ14.5/23, 20 ml
  • Type:
    For simultaneous distillation from 12×20 ml cylindrical flasks with 24/40 NS Joint