Spider evaporators
Spider with evaporating flasks, SJ24/40, glass, 50 ml, For simultaneous distillation from 5×50 ml evaporating flasks with 24/40 NS Joint
BUCH011574
: BUCHI
Spider evaporators are ideal when users need to concentrate several samples under the same conditions.
- Models for distillation with evaporating or cylindrical flasks available
- Parallel distillation saves time
- Description:Spider with evaporating flasks, SJ24/40, glass, 50 ml
- Type:For simultaneous distillation from 5×50 ml evaporating flasks with 24/40 NS Joint