Evaporating flasks
3000 ml, 24/40 NS, uncoated
N° de catalogue BUCH008767
Fournisseur: BUCHI
Aller à :
Product Details & Documents
Borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® rotary evaporators. Centre neck. Pear-shaped with ground joint socket or EL-flange.
Spécifications
- Socket size:24/40 NS
- Type:uncoated
- Capacity:3000 ml
- Seq:23