Orders LinkContactUsLinkComponent
Microplates, 384-well
Microplate, PP, flat bottom, 384-well, with fixed glass inserts, chemically inert, ~250 µl
Catalog # 391-1971
Supplier:   HIRSCHMANN
Microplates, 384-well
Catalog # 391-1971
Supplier:  HIRSCHMANN
Microplates, 384-well
Microplates, 384-well
Microplate, PP, flat bottom, 384-well, with fixed glass inserts, chemically inert, ~250 µl
Price
262,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Highly pure, chemically inert aluminium plate for individual glass inserts. Translucent plastic cover included.
Specifications
  • Description:
    Microplate, PP, flat bottom, 384-well, with fixed glass inserts, chemically inert
  • Well volume:
    ~250 µl
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Glass inserts for 384 well plates
    Glass inserts for 384 well plates
    HIRS9240584
    Log in for availability
    1 * 384 ST - 118,00 €