Order Entry
Belgium
Orders LinkContactUsLinkComponent
Gas sampling tubes

Gas sampling tubes

150 ml, without threaded bleed connection
Catalog # 201-9111
Supplier:   Lenz Laborglas
Gas sampling tubes
Catalog # 201-9111
Supplier:  Lenz Laborglas
Gas sampling tubes
Gas sampling tubes

150 ml, without threaded bleed connection

Price
93,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Duran® clear borosilicate glass.

  • Excellent thermal and chemical resistance
  • Connection on one side with capillary tube (internal Ø: 2,7 mm, external Ø: 8,0 mm) and on the other side with tube connector (external Ø: 8,0 mm)
  • With white labelling area

Gas collection tubes for removing, collecting and transporting gas samples. Both sides with a 3 NS/14 standard ground one-way stop cock.

Certifications: DIN 12473-1

Documents

Specifications
  • Type:
    without threaded bleed connection
  • Capacity:
    150 ml