Orders LinkContactUsLinkComponent
Extractor thimbles for solids
Extractor thimbles for solids
100 ml, 45/40 NS
Catalog # 533-2151
Supplier:   Lenz Laborglas
Extractor thimbles for solids
Catalog # 533-2151
Supplier:  Lenz Laborglas
Extractor thimbles for solids
Extractor thimbles for solids
100 ml, 45/40 NS
Price
119,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For glass inserts for solids.
Documents
Specifications
  • Extraction cap.:
    100 ml
  • Socket size:
    45/40 NS
  • Seq:
    02