Orders LinkContactUsLinkComponent
Multiple adapters
29/32 NS, 29/32 NS
  201-2217
Multiple adapters
  201-2217
Multiple adapters
Multiple adapters
29/32 NS, 29/32 NS
42,90 €
1 * 1 ST
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With one standard ground NS14/23 or NS 29/32 socket and two standard ground NS 14/23 or NS 29/32 cons. Side neck descending angle of 75°.
  • Cone size:
    29/32 NS
  • Socket size:
    29/32 NS