Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cylindrical separating funnels

Cylindrical separating funnels

50 ml, 29/32 NS, 1 ml
Catalog # 527-1101
Supplier:   Lenz Laborglas
Cylindrical separating funnels
Catalog # 527-1101
Supplier:  Lenz Laborglas
Cylindrical separating funnels
Cylindrical separating funnels

50 ml, 29/32 NS, 1 ml

Price
74,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass, with NS-PE stopper.

With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. With graduation.

Documents

Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    50 ml
  • Division:
    1 ml