Orders LinkContactUsLinkComponent
Stopcocks, one-way
8,0** mm, 24,0 NS
  227-3139
Stopcocks, one-way
  227-3139
Stopcocks, one-way
Stopcocks, one-way
8,0** mm, 24,0 NS
82,70 €
1 * 1 ST
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Replaceable NS keys
  • Screw thread retaining device
  • Leakage rate compliant with DIN 12540-1
Straight bore.
* with capillary side arms
** with rubber retaining device.
 : DIN 12554.
  • Hole Ø:
    8,0** mm
  • Seq:
    09
  • Spigot:
    24,0 NS