Orders LinkContactUsLinkComponent
Stopcocks, single-way, DURAN®
Stopcocks, single-way, DURAN®
2 mm, 12,5 NS
Catalog # 201-0383
Supplier:   DWK Life Sciences
Stopcocks, single-way, DURAN®
Catalog # 201-0383
Supplier:  DWK Life Sciences
Stopcocks, single-way, DURAN®
Stopcocks, single-way, DURAN®
2 mm, 12,5 NS
Price
1 320,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
  • Replaceable NS keys
  • Screw thread retaining device
Straight bore.
Specifications
  • Hole Ø:
    2 mm
  • Seq:
    18
  • Spigot:
    12,5 NS