Orders LinkContactUsLinkComponent
ORP electrode solutions
ORP electrode solutions
ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 640 mV, 60 Ampoul
Catalog # SCOI285138381
Supplier:   SI Analytics
ORP electrode solutions
Catalog # SCOI285138381
Supplier:  SI Analytics
CAS Number:  
ORP electrode solutions
ORP electrode solutions
ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 640 mV, 60 Ampoul
CAS Number:  
Price
224,00 €
Log in for availability
1 * 60 Ampul
Product Details & Documents
ORP electrode solutions are used as cleaning solutions for combination electrodes and reference electrodes.
Specifications
Specifications
  • Pk:
    60 Ampoul
  • Description:
    ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 640 mV