Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solutions

Type L4691, 10,01, ±0,010, 25 °C, 60 Ampoul
  SCOI285138205
Buffer solutions
  SCOI285138205
 :  
Buffer solutions
Buffer solutions

Type L4691, 10,01, ±0,010, 25 °C, 60 Ampoul

 :  
 
224,00 €
   
1 * 60 Ampul
 

Buffer solutions intended for extremely precise calibration.

  • Highest measurement reliability
  • No preservative agents
  • Hermetically sealed in the glass ampoule
  • Sterilised with hot steam

 : According to DIN 19 266.

 

 

  • Pk:
    60 Ampoul
  • Reference temperature:
    25 °C
  • Description:
    Type L4691
  • pH accuracy:
    ±0,010
  • pH value:
    10,01