Order Entry
Belgium
Orders LinkContactUsLinkComponent
ORP electrode solutions

ORP electrode solutions

ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 470 mV, 1 L
Catalog # 662-6171
Supplier:   SI Analytics
ORP electrode solutions
Catalog # 662-6171
Supplier:  SI Analytics
CAS Number:  
ORP electrode solutions
ORP electrode solutions

ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 470 mV, 1 L

CAS Number:  
Price
77,80 €
Log in for availability
1 * 1 L
Product Details & Documents

ORP electrode solutions are used as cleaning solutions for combination electrodes and reference electrodes.

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 470 mV

Specifications