Order Entry
Belgium
Orders LinkContactUsLinkComponent
Electrode Plug Cable Combinations

Electrode Plug Cable Combinations

Plug cable combinations, electrode socket, 10 m two core cable up to 80 °C, free end, 10 m
Catalog # SCOI285124708
Supplier:   SI Analytics
Electrode Plug Cable Combinations
Catalog # SCOI285124708
Supplier:  SI Analytics
Electrode Plug Cable Combinations
Electrode Plug Cable Combinations

Plug cable combinations, electrode socket, 10 m two core cable up to 80 °C, free end, 10 m

Price
99,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Electrode cables along with electrode socket, 1 m two core cable up to 80 °C, for resistance thermometer and 2-pole-conductivity cell without temperature sensor. Available with different cable length.

Specifications
  • Description:
    Plug cable combinations, electrode socket, 10 m two core cable up to 80 °C, free end
  • Length:
    10 m
  • Seq:
    04