Order Entry
Belgium
Orders LinkContactUsLinkComponent
Electrode Plug Cable Combinations

Electrode Plug Cable Combinations

Plug cable combinations, electrode socket, 1 m two core cable up to 80 °C, free end, 1 m
Catalog # SCOI285124613
Supplier:   SI Analytics
Electrode Plug Cable Combinations
Catalog # SCOI285124613
Supplier:  SI Analytics
Electrode Plug Cable Combinations
Electrode Plug Cable Combinations

Plug cable combinations, electrode socket, 1 m two core cable up to 80 °C, free end, 1 m

Price
50,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Electrode cables along with electrode socket, 1 m two core cable up to 80 °C, for resistance thermometer and 2-pole-conductivity cell without temperature sensor. Available with different cable length.

Specifications
  • Description:
    Plug cable combinations, electrode socket, 1 m two core cable up to 80 °C, free end
  • Length:
    1 m
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.