Order Entry
Belgium
Orders LinkContactUsLinkComponent
Electrode Cables

Electrode Cables

Electrode cable with 4 mm banana connector, 1 m
Catalog # 662-6119
Supplier:   SI Analytics
Electrode Cables
Catalog # 662-6119
Supplier:  SI Analytics
Electrode Cables
Electrode Cables

Electrode cable with 4 mm banana connector, 1 m

Price
41,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

Electrode cables with a range of connectors to fit most pH meters.

Specifications
  • Description:
    Electrode cable with 4 mm banana connector
  • Length:
    1 m