Order Entry
Belgium
Orders LinkContactUsLinkComponent
Filter discs, DURAN®

Filter discs, DURAN®

1, 40 mm, 3,8 mm
  SCOT251540109
Filter discs, DURAN®
  SCOT251540109
Filter discs, DURAN®
Filter discs, DURAN®

1, 40 mm, 3,8 mm

 
94,40 €
   
1 * 10 ST
 

Sintered glass filters have neutral properties with unfused edge and untreated surfaces. The hard sintering process makes the material resilient.

  • Available in intermediate sizes, alternative dimensions, alternate porosities and bi-planar design

Made from clear DURAN® borosilicate glass 3,3.

 
  • Ø:
    40 mm
  • Porosity:
    1
  • Seq:
    24
  • Thickness:
    3,8 mm