Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Measuring cylinders tall form, without base, DURAN®
Measuring cylinders tall form, without base, DURAN®
25 ml
Catalog # 215-3625
Supplier:   DWK Life Sciences
Measuring cylinders tall form, without base, DURAN®
Catalog # 215-3625
Supplier:  DWK Life Sciences
Measuring cylinders tall form, without base, DURAN®
Measuring cylinders tall form, without base, DURAN®
25 ml
Price
70,60 €
Log in for availability
1 * 10 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Tall form
  • Flat bottom
  • With spout
  • Without base
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
  • Without foot
Specifications
  • Capacité:
    25 ml
  • Seq:
    02