Orders LinkContactUsLinkComponent
Gooseneck light guides
Gooseneck light guides
3-arm gooseneck light guide, Ø 4,5 mm, L=600 mm, KL 1500 HAL / KL 1500 compact / KL 1600 LED / 2500 LCD / KL 2500 LED
Catalog # 631-0023
Gooseneck light guides
Catalog # 631-0023
Gooseneck light guides
Gooseneck light guides
3-arm gooseneck light guide, Ø 4,5 mm, L=600 mm, KL 1500 HAL / KL 1500 compact / KL 1600 LED / 2500 LCD / KL 2500 LED
Price
478,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Fibre optic illumination for stereo microscopy. To be used with cold light sources.
In all its microscopy light guides, SCHOTT uses the new environmentally friendly glass optical fibers PURAVIS®. Not just the fibers themselves, but also the entire proprietary manufacturing process are lead free and avoid the use of arsenic and antimony. Due to superior physical and chemical properties, PURAVIS® glass optical fibers provide a high performance, superior longevity, improved transmission in the near UV and outstanding transmission of white light.
Specifications
  • Description:
    3-arm gooseneck light guide, Ø 4,5 mm, L=600 mm
  • Seq:
    310
  • For:
    KL 1500 HAL / KL 1500 compact / KL 1600 LED / 2500 LCD / KL 2500 LED