Burette stopcocks, straight
Burettes of 25 ml capacity
Catalog # BRND81937
Supplier: Brand
Jump to:
Product Details & Documents
Borosilicate glass 5,4, amber.
- Interchangeable key
- With glass or PTFE key
- Key with retention device
Stopcock housing with burette tip and key.
Specifications
- For:Burettes of 25 ml capacity
- Seq:03
- Cat. No.:BRND81937