Orders LinkContactUsLinkComponent
Burette stopcocks, lateral
Burette stopcocks, lateral
Stopcock with PTFE key, Burettes of 25 and 50 ml capacity
Catalog # BRND81927
Supplier:   Brand
Burette stopcocks, lateral
Catalog # BRND81927
Supplier:  Brand
Burette stopcocks, lateral
Burette stopcocks, lateral
Stopcock with PTFE key, Burettes of 25 and 50 ml capacity
Price
57,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 5,4, amber.
  • Interchangeable key
  • With glass or PTFE key
  • Key with retention device
Stopcock housing with burette tip and key.
Specifications
  • For:
    Burettes of 25 and 50 ml capacity
  • Description:
    Stopcock with PTFE key
  • Seq:
    03