Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Desiccator lids, with socket

Desicator lid, Hxø: 84x215 mm, With socket, Novus, 150 DN, 85 mm, 215 mm
  BRND65731
 :   Brand
Desiccator lids, with socket
  BRND65731
 :  Brand
Desiccator lids, with socket
Desiccator lids, with socket

Desicator lid, Hxø: 84x215 mm, With socket, Novus, 150 DN, 85 mm, 215 mm

 
131,00 €
   
1 * 1 ST
 

Dessicator lids with socket 24/29, made of borosilicate glass 3.3.

 
  • Ext.Ø:
    215 mm
  • Nominal size:
    150 DN
  • Type:
    With socket, Novus
  • Description:
    Desicator lid, Hxø: 84x215 mm
  • Height:
    85 mm
  • Seq:
    01