Orders LinkContactUsLinkComponent
Cavity slides
Cavity slides
Cavity slides, Clear, 1, 15 - 18 mm
Catalog # BRND475505
Supplier:   Brand
Cavity slides
Catalog # BRND475505
Supplier:  Brand
Cavity slides
Cavity slides
Cavity slides, Clear, 1, 15 - 18 mm
Price
167,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
Cavity slides in hydrolytic class 3 glass.
  • Ground edges
Specifications
  • No. of wells:
    1
  • Colour:
    Clear
  • Description:
    Cavity slides
  • Seq:
    01
  • Well Ø:
    15 - 18 mm