Orders LinkContactUsLinkComponent
Microscopy slides
Microscopy slides
Slides with cut edges and twinfrosted end, White/clear, 76×26 mm
Catalog # BRND474702
Supplier:   Brand
Microscopy slides
Catalog # BRND474702
Supplier:  Brand
Microscopy slides
Microscopy slides
Slides with cut edges and twinfrosted end, White/clear, 76×26 mm
Price
326,00 €
Log in for availability
1 * 2.500 ST
Product Details & Documents
Microscopy slides with ground edges to reduce the risk of injury.
Specifications
  • Colour:
    White/clear
  • Description:
    Slides with cut edges and twinfrosted end
  • L×W:
    76×26 mm