Watch glasses
Watch glass, soda-lime glass, with fire-polished edges, 120 mm
Catalog # BRND150060
Supplier: Brand
Jump to:
Product Details & Documents
Soda-lime glass.
- Good chemical and physical properties
- Use in applications that do not involve high thermal stresses
- Quick and easy to clean
With fire-polished edges. Stress-reduced to minimise risk of shattering.
Certifications: DIN 12341.
Specifications
- Ø:120 mm
- Description:Watch glass, soda-lime glass, with fire-polished edges
- Seq:08
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.