Orders LinkContactUsLinkComponent
Watch glasses
Watch glasses
Watch glass, soda-lime glass, with fire-polished edges, 40 mm
Catalog # BRND150010
Supplier:   Brand
Watch glasses
Catalog # BRND150010
Supplier:  Brand
Watch glasses
Watch glasses
Watch glass, soda-lime glass, with fire-polished edges, 40 mm
Price
30,60 €
Log in for availability
1 * 10 ST
Product Details & Documents
Soda-lime glass.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean
With fire-polished edges. Stress-reduced to minimise risk of shattering.
Certifications: DIN 12341.
Specifications
  • Ø:
    40 mm
  • Description:
    Watch glass, soda-lime glass, with fire-polished edges
  • Seq:
    01