Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mini Coolers

Mini Coolers

Mini cooler, maintains the Temperature at −70 °C during 45 Minutes, 0,5 to 2,0 ml tubes, 12, White
Catalog # 479-1243
Supplier:   Brand
Mini Coolers
Catalog # 479-1243
Supplier:  Brand
Mini Coolers
Mini Coolers

Mini cooler, maintains the Temperature at −70 °C during 45 Minutes, 0,5 to 2,0 ml tubes, 12, White

Price
126,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PC, yellow, with clear lid. Maintains temperature of 0 °C for 60 min.

  • Durable polycarbonate filled with non-toxic gel
  • Mini coolers hold twelve 0,5 to 2,0 ml tubes

Mini coolers are designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench.

Specifications
  • For:
    0,5 to 2,0 ml tubes
  • No. of Holes:
    12
  • Colour:
    White
  • Description:
    Mini cooler, maintains the Temperature at −70 °C during 45 Minutes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.