Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mini Coolers

Mini Coolers

Mini cooler, maintains the Temperature at −20 °C during 60 Minutes, 0,5 to 2,0 ml tubes, 12, Yellow
Catalog # 479-1242
Supplier:   Brand
Mini Coolers
Catalog # 479-1242
Supplier:  Brand
Mini Coolers
Mini Coolers

Mini cooler, maintains the Temperature at −20 °C during 60 Minutes, 0,5 to 2,0 ml tubes, 12, Yellow

Price
126,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PC, yellow, with clear lid. Maintains temperature of 0 °C for 60 min.

  • Durable polycarbonate filled with non-toxic gel
  • Mini coolers hold twelve 0,5 to 2,0 ml tubes

Mini coolers are designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench.

Specifications
  • For:
    0,5 to 2,0 ml tubes
  • No. of Holes:
    12
  • Colour:
    Yellow
  • Description:
    Mini cooler, maintains the Temperature at −20 °C during 60 Minutes