Mini Coolers
Mini Cooler, maintains the Temperature at 0 °C during 60 Minutes, 0,5 to 2,0 ml tubes, 12, Red
Catalog # 479-1241
Supplier: Brand
Jump to:
Product Details & Documents
PC, yellow, with clear lid. Maintains temperature of 0 °C for 60 min.
- Durable polycarbonate filled with non-toxic gel
- Mini coolers hold twelve 0,5 to 2,0 ml tubes
Mini coolers are designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench.
Specifications
- For:0,5 to 2,0 ml tubes
- No. of Holes:12
- Colour:Red
- Description:Mini Cooler, maintains the Temperature at 0 °C during 60 Minutes



