Orders LinkContactUsLinkComponent
Accessories for Spectroquant® NOVA
Accessories for Spectroquant® NOVA
Software for Windows (in English and German) for the data transfer, NOVA photometers Spectroquant®
Catalog # 1.14964.0001
Supplier:   Merck
Accessories for Spectroquant® NOVA
Catalog # 1.14964.0001
Supplier:  Merck
Accessories for Spectroquant® NOVA
Software for Windows (in English and German) for the data transfer, NOVA photometers Spectroquant®
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
To document the analysis values, either a standard computer or a printer can be connected.
  • The Multi-Agate II software can control data acquisition or simply record recorded analysis data
  • The data records are archived here as raw data or imported into further programs such as Excel
Connectable photometers: SQ 118, SQ 200, SQ 300, Spectroquant® NOVA 30, Spectroquant® NOVA 60, Spectroquant® NOVA 400.
Specifications
  • Description:
    Software for Windows (in English and German) for the data transfer
  • Seq:
    905
  • For:
    NOVA photometers Spectroquant®