Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

n-Pentane ≥95%, TECHNICAL

Metal drum, 25 L
Catalog # 26192.463
Flammable
Health
Irritant
Environment
Supplier:   VWR Chemicals
n-Pentane ≥95%, TECHNICAL
Catalog # 26192.463
Supplier:  VWR Chemicals
CAS Number:  109-66-0
n-Pentane ≥95%, TECHNICAL
n-Pentane ≥95%, TECHNICAL

Metal drum, 25 L

CAS Number:  109-66-0
Price
437,00 €
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Pentane
  • CAS Number:109-66-0
  • Formula:H₃C(CH₂)₃CH₃
  • MDL Number:MFCD00009498
  • Molecular Weight:72.15 g/mol
  • Merck Index:13,07193
  • Boiling Pt:35…36 °C (1013 hPa)
  • Melting Pt:–130 °C
  • Density:0.63 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • Storage Temperature:Ambient
  • EINECS:203-692-4
  • UN:1265
  • ADR:3,II
  • REACH:01-2119459286-30
Specifications

Specification Test Results

  • Assay:
    Min. 95 %
  • Boiling point:
    35 - 37 °C

Specifications

  • Pack type:
    Metal drum
  • Pk:
    25 L

Specifications