Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH-Fix pH Indicator Test Strips

pH-Fix pH Indicator Test Strips

pH-Fix, 100 test strips with colour scale, 0,3-2,3, Classic flat box , 65×10×90 mm, 32,4 g
Catalog # 662-2343
Supplier:   MACHEREY-NAGEL
pH-Fix pH Indicator Test Strips
Catalog # 662-2343
Supplier:  MACHEREY-NAGEL
pH-Fix pH Indicator Test Strips
pH-Fix pH Indicator Test Strips

pH-Fix, 100 test strips with colour scale, 0,3-2,3, Classic flat box , 65×10×90 mm, 32,4 g

Price
16,10 €
Log in for availability
1 * 100 ST
Product Details & Documents

pH-Fix indicator test strips allow fast pH testing directly at the point of interest, and are suitable for untrained users as well as analytical professionals.

  • No accessories needed - always ready to use
  • Brilliant colour chart for precise readings
  • Long handle for sufficient hand - sample - distance
  • Non bleeding - no contamination of sample solutions

The indicator dyes used for pH-Fix test strips are chemically bound to the test pads to prevent bleeding and, therefore, contamination of the sample. Instructions for use according to IVD Directive 98/79/EC - The instructions for use according to IVD Directive 98/79/EC can be requested by providing the article number and LOT number at the email address [email protected].

Caution: No hazardous material.

Specifications
  • pH range:
    0,3-2,3
  • Description:
    pH-Fix, 100 test strips with colour scale
  • Seq:
    4
  • Pack type:
    Classic flat box , 65×10×90 mm, 32,4 g
  • Cat. No.:
    662-2343
  • Shelf life:
    4 Year(s)