Order Entry
Belgium
Orders LinkContactUsLinkComponent
Test fluids for viscometers

Test fluids for viscometers

E7, 5, 100 mL
Catalog # HAAK082-5042
Supplier:   HAAKE
Test fluids for viscometers
Catalog # HAAK082-5042
Supplier:  HAAKE
CAS Number:  
Test fluids for viscometers

E7, 5, 100 mL

CAS Number:  
Price
209,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Test and certificated fluids for viscometery - mesurement temperature at 20 °C unless indicated.

Caution: * Measurement temperature 25 °C

Documents

Specifications

Specifications

  • Pk:
    100 mL
  • Type:
    E7
  • Viscosity:
    5

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.