Transport containers
Slide mailers, PP, 82×17×29 mm, 5
Fournisseur: DELTALAB
Aller à :
Product Details & Documents
Designed for slide transport and protection.
- Made of polypropylene
- Easy snap-lock
- Not indicated for liquid samples
Compartment shape ensures tight fit: no risk of damaging slides and to send by mail, recommend to use special envelopes.
Documents
Certificats
Rechercher par numéro de lotSpécifications
- No. of slides:5
- Description:Slide mailers
- Material:PP
- Seq:05
- W×D×H:82×17×29 mm