Orders LinkContactUsLinkComponent
Slide boxes
Slide boxes
White, PS, 25, 105×88×32 mm
Catalog # 631-9710
Supplier:   DELTALAB
Slide boxes
Catalog # 631-9710
Supplier:  DELTALAB
Slide boxes
Slide boxes
White, PS, 25, 105×88×32 mm
Price
12,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stackable items to gain space on the bench.
  • Translucent lid to see if the box is empty or not
  • No numbering of the slots
Documents
Specifications
  • No. of slides:
    25
  • Colour:
    White
  • Material:
    PS
  • Seq:
    01
  • W×D×H:
    105×88×32 mm