Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue processing cassettes, polyacetylene

Tissue processing cassettes, polyacetylene

Red
Catalogus # 631-9643
Leverancier:   DELTALAB
Tissue processing cassettes, polyacetylene
Catalogus # 631-9643
Leverancier:  DELTALAB
Tissue processing cassettes, polyacetylene

Red

Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 1.000 ST
Product Details & Documents

These cassettes are made of polyacetyle.

  • With 34 round holes of 2 mm
  • External dimensions: 40×28×6 mm
  • Can be written on

They are resistant to the solvents used in histological processes.

Documents

Specificaties
  • Colour:
    Red
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.