Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions, colour coded, Fluka™

Buffer solutions, colour coded, Fluka™

4,00, Red, 20 °C, Plastic bottle, 0,5 L, 500 mL
Catalog # 33665-500ML
Supplier:   Honeywell Chemicals
Buffer solutions, colour coded, Fluka™
Catalog # 33665-500ML
CAS Number:  
Buffer solutions, colour coded, Fluka™

4,00, Red, 20 °C, Plastic bottle, 0,5 L, 500 mL

CAS Number:  
Price
42,40 €
Log in for availability
1 * 500 mL
Product Details & Documents

These buffer solutions are colour-coded for easy identification and the pH 4 and 7 version have fungicide additive.

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    20 °C
  • Colour:
    Red
  • Pk Info:
    0,5 L
  • pH value:
    4,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.