Buffer solutions, colour coded, Fluka™
4,00, Red, 20 °C, VOLPAK container, 10 L, 10 L
Catalog # 33665-10L-VP
Supplier: Honeywell Chemicals
Buffer solutions, colour coded, Fluka™
Catalog # 33665-10L-VP
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
These buffer solutions are colour-coded for easy identification and the pH 4 and 7 version have fungicide additive.
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:VOLPAK container
- Pk:10 L
- Reference temperature:20 °C
- Colour:Red
- Pk Info:10 L
- pH value:4,00



